VWR Ovens and Incubators
Jaw Dropping Deals! Min. 20% OFF across various categories.
Natural convection lab ovens – DRY-Line PRIME
The DRY-Line® drying ovens are designed for all standard drying and tempering tasks as well as glassware drying. They have a temperature range between 50 to 300°C and are intended for use in biological, chemical, medical, pharmaceutical and physical laboratories.
- Excellent temperature control
- Ovens based on natural convection for multiple application
- Over temperature protection to meet your safety requirements
Forced convection lab ovens – VENTI-Line PRIME
VENTI-Line® drying ovens are designed for all standard drying and tempering tasks as well as glassware sterilization. The Forced convection feature ensures excellent temperature control and uniformity in the Chamber, suitable for a wide range of application.
- Forced convection for excellent temperature uniformity
- Fan speed control to meet all your application needs
- Excellent temperature control for stringent requirements
Cooled Incubators – INCU-Line Basic and Premium Series
These compressor-cooled incubators with forced air convection provide a temperature range between 3 to 70°C. Featuring a stainless steel (DIN 1.4016) inner chamber and powder coated sheet exterior. These models have an intuitive menu, USB interface for data download to USB stick, and can be connected to LAN network.
- Forced air convection for excellent temperature uniformity
- All-New 4.3” coloured touch screen display
- 5-segment temperature-time profile & adjustable ramp times to meet any programming requirements
Cooled Incubators – INCU-Line ILPR Premium Series
Cooling incubators with peltier technology feature excellent temperature stability and uniformity. They are energy efficient and environmentally friendly, with a low noise level, and they work almost vibration-free as no compressor is installed.
- Fast temperature control with peltier technology
- Excellent chemical resistance with stainless steel interior and exterior
- Suitable for GLP environments
Natural lab Incubators – INCU-Line PRIME
INCU-Line® microbiological incubators are designed to cultivate organisms at homogeneous temperatures. They have a temperature range of 5°C above ambient to 100°C and can, for instance, be used for microbiological tests, germ count determination and ageing test / Shelf-Lift testing / heated storage.
- Natural convection for your basic incubation needs
- Inner & outer doors to prevent heat loss
- Powder coated housing with stainless steel interior for chemical protection
VIEW other ranges at VWR Reliable Laboratory Equipment
Click on "Request a Quote" to enjoy our exclusive deals now!
Description | UOM | VWR Catalog Number |
VWR DRY-Line® 56 Prime | 1 * 1 items | VWRI466-0600 |
VWR DRY-Line® 112 Prime | 1 * 1 items | VWRI466-0601 |
VWR DRY-Line® 180 Prime | 1 * 1 items | VWRI466-0602 |
VWR VENTI-Line® 56 Prime | 1 * 1 items | VWRI466-0603 |
VWR VENTI-Line® 112 Prime | 1 * 1 items | VWRI466-0604 |
VWR VENTI-Line® 180 Prime | 1 * 1 items | VWRI466-0605 |
VWR INCU-Line® IL 68R cooled incubator | 1 * 1 items | VWRI390-1341 |
VWR INCU-Line® IL 150R cooled incubator | 1 * 1 items | VWRI390-1342 |
VWR INCU-Line® IL 250R cooled incubator | 1 * 1 items | VWRI390-1343 |
VWR INCU-Line® IL 150R cooled incubator PREMIUM | 1 * 1 items | VWRI390-1338 |
VWR INCU-Line® IL 250R cooled incubator PREMIUM | 1 * 1 items | VWRI390-1339 |
VWR INCU-Line® IL 56PR Peltier cooled incubator | 1 * 1 items | VWRI390-1351 |
VWR INCU-Line® IL 112PR Peltier cooled incubator | 1 * 1 items | VWRI390-1352 |
VWR INCU-Line® IL 240PR Peltier cooled incubator | 1 * 1 items | VWRI390-1353 |
VWR INCU-Line® IL 56 Prime incubator | 1 * 1 items | VWRI390-1346 |
VWR INCU-Line® IL 112 Prime incubator | 1 * 1 items | VWRI390-1349 |
VWR INCU-Line® IL 180 Prime incubator | 1 * 1 items | VWRI390-1350 |