Memmert Ovens and Incubators
Sizzling Hot Deals! Up to 27% off. Don’t miss it.*T&Cs Apply!
Incubators, Generation 2012, IF/IFplus (forced convection)
Incubators, Generation 2012, IN/INplus (natural convection)
The Generation 2012 range has natural and forced convection models with a temperature range up to +80 °C. There is a choice of two different displays, SingleDISPLAY or TwinDISPLAY depending on the application.
- SingleDISPLAY models, ideal for standard applications
- ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, program time, time zones and daylight-saving time
Cooled Incubators with Advanced Peltier Technology, IPP and IPPeco series
Sets eco standards for cultivation below room temperature: Unmatched energy efficiency, excellent heat-up, cool-down and recovery times.
- Maximum energy efficiency compared to cooled incubators with compressor
- Completely homogenous distribution of temperature
Certifications: Standard units are safety-approved and bear the EAC test marks.
Ordering information: Supplied without accessories.
Heating and drying ovens, Generation 2012 Universal UF/UFplus (forced convection)
Heating and drying ovens, Generation 2012 Universal UN/UNplus (natural convection)
The Generation 2012 range includes forced convection models with a temperature range up to +300 °C. There is a choice of two different displays, SingleDISPLAY or TwinDISPLAY depending on the application.
- SingleDISPLAY models, ideal for standard applications
- ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, program time, time zones and daylight saving time
Accessories information: For details of the range of shelves and grids, please visit vwr.com or contact your local Avantor sales office. *Applicable to Incubators, Generation 2012, IF/IFplus too
Ordering information: Supplied without shelves, please order required shelves, stainless steel grids or trays separately. Range of accessories and factory fitted options such as access ports, interior sockets, lighting and interior door are available on request. Please contact Avantor for further details prior to ordering as these cannot be retro-fitted. *Applicable to Incubators, Generation 2012, IF/IFplus too
Description | UOM | VWR Catalog Number |
---|---|---|
IF110 with SingleDISPLAY | 1 * 1 items | MEMMIF110 |
IF55 with SingleDISPLAY | 1 * 1 items | MEMMIF55 |
IF75 with SingleDISPLAY | 1 * 1 items | MEMMIF75 |
IN110 with SingleDISPLAY | 1 * 1 items | MEMMIN110 |
IN160 with SingleDISPLAY | 1 * 1 items | MEMMIN160 |
IN260 with SingleDISPLAY | 1 * 1 items | MEMMIN260 |
IN30 with SingleDISPLAY | 1 * 1 items | MEMMIN30 |
IN55 with SingleDISPLAY | 1 * 1 items | MEMMIN55 |
IN75 with SingleDISPLAY | 1 * 1 items | MEMMIN75 |
IPP110eco | 1 * 1 items | MEMMIPP110ECO |
UF110 with SingleDISPLAY | 1 * 1 items | MEMMUF110 |
UF160 with SingleDISPLAY | 1 * 1 items | MEMMUF160 |
UF30 with SingleDISPLAY | 1 * 1 items | MEMMUF30 |
UF55 with SingleDISPLAY | 1 * 1 items | MEMMUF55 |
UN110 with SingleDISPLAY | 1 * 1 items | MEMMUN110 |
UN55 with SingleDISPLAY | 1 * 1 items | MEMMUN55 |
UN75 with SingleDISPLAY | 1 * 1 items | MEMMUN75 |
*Terms and Conditions:
- Promotion valid from now till 31 December 2023
- VWR standard Terms and Conditions apply