IKA Instruments Deal
Stirrers, Shaker, Disperser and Viscometer on Sale. 25% OFF! While stocks last. *T&Cs Apply.
Electronic overhead stirrers, EUROSTAR 60 digital, EUROSTAR 60 control, EUROSTAR 100 digital and EUROSTAR 100 control
Powerful laboratory stirrers for quantities up to 40 or 100 litres (H₂O). Units automatically adjust the speed through microprocessor-controlled technology within the speed range of 0/30 to 2000 min⁻¹ (EUROSTAR 60 models) or 0/30 to 1300 min⁻¹ (EUROSTAR 100 models). Safety circuits ensure automatic cut-off in anti-stall or overload conditions. The actual shaft and pre-set speed are constantly monitored, and variations adjusted automatically. This guarantees a constant speed even with changes in sample viscosities.
- Digital speed display
- Infinitely adjustable speed
Certifications: Protection class according to DIN EN 60529: IP 40.
Package Deals for below Eurostar and RW:
- EUROSTAR 60 Digital Package, consists of IKAA4446002 – Eurostar 60 digital, IKAA741000 – R 1342, IKAA2657700 - R 182, IKAA3160100 - R 1826, IKAA3008600 – RH 3 (1 * 1 KIT)
- STIRRER OVERH EUROSTAR 20 DIG.PACK UK, consists of IKAA741000 – R 1342, IKAA2657700 – R 182, IKAA3160100 – R 1826, IKAA4442002 – EUROSTAR 20 Digital (1 * 1 items)
- RW 20 Digital Package (incl. R 1826, R 182, R 1342), including Plate stand, Propeller stirrer and Boss head clamp (1 * 1 items)
The bestseller in the laboratory:
- With digital display
- Robust, slimline, ergonomic design
- Technical improvements on the trusted RW 20 series designs
- With constant power-drive
- Two speed ranges for universal use from 60 - 2.000 rpm
- Push-through agitator shafts (only when stationary)
- For stirring quantities of up to 20 l (H2O)
Orbital shakers, KS 260
Compact flat shakers with electronic adjustment of speed and timer. Control model has RS232 analogue interface, integrated end point positioning for automated robot controlled sampling and all functions can be controlled and documented with optional labworldsoft® software.
- Continuous or timer operation; basic model from 5 to 50 min, control model 0 to 599 min
- Can be used at 5 to 50 °C (80% relative humidity), in an incubator or cold room
Certifications: IP protection class according to DIN EN 60529: IP 21
Dispenser, T25 DIGITAL ULTRA-TURRAX PACKAGE-UK PLUG
High-performance dispersing instrument for volumes from 1 – 2000 ml (H2O) with digital speed display. It offers a wide speed range from 3000 – 25,000 rpm that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements adds versatility. Applications ranges from homogenizing waste water samples, use in laboratory reactors, dispersion tasks under vacuum / pressure and sample preparation in medical research.
- Digital speed display
- Electronic speed control
- Electronic overload protection
- Stainless steel dispersing elements can be cleaned quickly and easily
- Plastic disposable dispersing elements are available in two sizes
- Error code display
- Quiet operation
Rotational viscometers, ROTAVISC
The ROTAVISC series determines the viscosity of liquids in all areas of application from the laboratory to quality control.
- Easy to operate, highest measurement accuracy and stepless speed
- TFT display for an intuitive menu guidance
Certifications: IP 40 protection class according to DIN EN 60529.
IKA's range of instruments is the culmination of years of innovation. The Eurostar and RW overhead stirrers are suited for a wide range of samples for your lab, each model is carefully manufactured to suit your stirring needs. If you are in need of a disperser, the IKA Ultra-Turrax range is a disperser suited for different samples types and ensures a homogenous product. The KS 260 Orbital shaker is a top of the line shaker and it has multiple attachments for various samples. Finally, the IKA viscometer is a benchtop unit that provides crucial information for your sample, it measures the viscosity quickly and accurately without using much of your bench space.
Description | UOM | VWR Catalog Number |
STIRRER OVERHEAD EUROSTAR 100 DIGIT UK | 1 * 1 items | IKAA4238102 |
STIRRER OVERHEAD EUROSTAR 60 CONTROL | 1 * 1 items | IKAA4440000 |
STIRRER OVERHEAD EUROSTAR 60 CONT S 2 UK | 1 * 1 items | IKAA4440002 |
EUROSTAR 60 DIGITAL PACKAGE | 1 * 1 KIT | IKAA10000354 |
STIRRER OVERH EUROSTAR 20 DIG.PACK UK | 1 * 1 items | IKAA9025102 |
STIRRER RW 20 DIGITAL PACKAGE UK-PLUG | 1 * 1 items | IKAA9011602 |
SHAKER ORBITAL MODEL KS260 BASIC - UK | 1 * 1 items | IKAA2980202 |
T25 DIGITAL ULTRA-TURRAX PACKAGE-UK PLUG | 1 * 1 items | IKAA9024202 |
VISCOSIMETER ROTAVISC LO-VI COMPLETE ROT | 1 * 1 items | IKAA25000310 |
*Terms and Conditions:
|