QuantaBio qScript Ultra SuperMix and Ultra Flex
New Product Launch! qScript Ultra Flex SuperMix Kit and qScript Ultra Flex Kit, from Quantabio, for faster, longer and tougher next generation cDNA synthesis with flexible priming. Get your free sample today! *T&Cs Apply!
qScript® Ultra SuperMix
- Enhanced Thermostability: Engineered for Reverse Transcriptase activity up to 65°C, overcoming challenging secondary structures of RNA
- Faster Transcription & Ease of Use: Single tube master mix generates cDNA in 10 minutes. Simply add your RNA template
- Maximum Yield and Sensitivity: Reproducible, low abundance detection over a linear dynamic range from 1 pg – 2.5 ug
- Balanced Coverage of Long RNATranscripts: Unbiased transcriptome representation in the cDNA product for longer gene expression analysis.
- Ultimate Inhibitor Resistance: Overcome a wide array of PCR inhibitors (salt, heparin, hematin, etc.) in difficult samples
qScript Ultra SuperMix is a highly stabilized, efficient and easy-to-use single tube master mix for the synthesis of first-strand cDNA to reverse transcribe RNA to cDNA. A key component is a novel, state-of-the-art, RNase H deficient reverse transcriptase that was engineered for improved thermostability, velocity, processivity, and resistance to many common inhibitors. qScript Ultra SuperMix contains all required components for first-strand cDNA synthesis except RNA template and is directly compatible with downstream 2-step RT-qPCR or RT-PCR procedures.
qScript® Ultra Flex kit (25R)
qScript® Ultra Flex kit (100R)
Faster, longer, tougher next gen cDNA synthesis with flexible priming.
- Faster, longer, tougher next gen cDNA synthesis with flexible priming.
- Enhanced thermostability – Engineered for RT activity up to 65 °C, overcoming challenging secondary structures
- Superior speed and ease of use – 10 minute reaction time with fewer pipetting steps
- Maximum yield and sensitivity – Improved cDNA yield with limiting or compromised samples
- Ultimate inhibitor resistance – Overcome a wide array of PCR inhibitors (salt, heparin, hematin, etc.)
- Longer transcripts – Ability to reverse transcribe single targets up to 20 kb
qScript Ultra Flex Kit is easy-to-use and highly efficient for the synthesis of first-strand cDNA with flexible priming methods including Oligo-dT, Random Hexamer and Gene Specific Priming to reverse transcribe RNA to cDNA. A key component is a novel, state-of-the-art, RNase H deficient reverse transcriptase that was engineered for improved thermostability, velocity, processivity, and resistance to many common reaction inhibitors. qScript Ultra Flex Kit contains all required components for first-strand cDNA synthesis except RNA template and gene specific primers. The cDNA product is directly compatible with downstream 2-step RT-qPCR or RT-PCR procedures.
qScript Ultra Flex Kit has an optimal reaction temperature of 55°C and a 10-min reaction time, significantly reducing time to result while working through challenging secondary RNA structures. The lower Cq values for the engineered qScript Ultra reverse transcriptase indicate more efficient cDNA synthesis at high temperatures, critical for progressing through high GC regions and regions of RNA secondary structure.
Request a Quote Request SampleDescription | UOM | VWR Catalog Number |
QSCRIPT ULTRA SUPERMIX (25R) | 1 * 25 Tests | QUNT95217-025 |
QSCRIPT ULTRA SUPERMIX (100R) | 1 * 100 Tests | QUNT95217-100 |
QSCRIPT ULTRA SUPERMIX (500R) | 1 * 500 Tests | QUNT95217-500 |
QSCRIPT ULTRA FLEX KIT (25R) | 1 * 25 Tests | QUNT95215-025 |
QSCRIPT ULTRA FLEX KIT (100R) | 1 * 100 Tests | QUNT95215-100 |
*Terms and Conditions:
- Promotion valid from now till 30th June 2023
- The Free sampling program is only for the smaller pack sizes, QUNT95217-025 and QUNT95215-025
- VWR standard Terms and Conditions apply