Avantor VWR Cryovials New Launch
For your cryopreservation applications. Up to 20% off! While stocks last. *T&Cs Apply.
Designed for freezing samples in all cryopreservation applications, these vials feature an internal thread with washer, or an external thread.
- Manufactured from polypropylene to withstand temperatures down to –196 °C
- Manufactured in cleanroom environment (Class 10,000)
- Free of RNase, DNase and pyrogens
- White graduations, write on area on vial for easy marking
- Internal vials have a washer for a secure seal
- 2D coded vial available in 2 ml self-standing
- Vials may be colour coded with inserts
- Sterile (gamma irradiated)
- Cap material: HDPE
Self standing/starfoot/round bottom vial option for user to choose based on the application.
Cryocoder/cap inserts are available in multiple colour options: Violet, white, blue, orange, lilac, red, tan, yellow, green, grey, pink, assorted (five colours).
Certifications: US FDA 21CRF, USP CLASS VI compliant Resin.
Caution: Store cryogenic vials only in the vapour phase above the liquefied gas. Always use appropriate safety equipment when removing vials from cryogenic storage.
Description | UOM | VWR Catalog Number |
CRYOVIAL EXT TRD SS 1.8/2 ML | 1 * 1000 items | VWRI210-000004 |
CRYOVIAL EXT TRD SS 3 ML | 1 * 1000 items | VWRI210-000006 |
CRYOVIAL EXT TRD SS 3.6/4 ML | 1 * 500 items | VWRI210-000009 |
CRYOVIAL EXT TRD SS 4.5/5 ML | 1 * 500 items | VWRI210-000010 |
CRYOVIAL EXT TRD RB 1.8/2 ML | 1 * 1000 items | VWRI210-000005 |
CRYOVIAL INT TRD SS 1.8/2 ML W/WASHER | 1 * 1000 items | VWRI210-000014 |
CRYOVIAL INT TRD RB 1.8/2 ML W/WASHER | 1 * 1000 items | VWRI210-000015 |
CRYOVIAL INT TRD RB 4.5/5 ML W/WASHER | 1 * 500 items | VWRI210-000021 |
CRYOVIAL 2D EXT TRD W/CAP SS 2ML | 1 * 1000 items | VWRI210-000026 |
CAP INSERT 5 COLORS | 1 * 500 items | VWRI210-000033 |
*Terms and Conditions:
- Promotion valid from now till 30 June 2024 *While Stocks Last
- VWR standard Terms and Conditions apply
- All pre-orders must be made within the promotional period
- Avantor VWR Cryovials New Launch is not applicable with other on-going promotions