KC Polaris Gloves New Launch
Kimtech™ Polaris™ Nitrile and Xtra Nitrile Gloves are built for science, made and designed for use in laboratories, life science applications, and non-sterile pharmaceutical manufacturing environments. Up to 14% OFF! While stocks last. *T&Cs Apply.
- High-end ambidextrous powder-free nitrile gloves deliver seamless protection whenever and wherever it is needed
- Optimized nitrile formulation (acrylonitrile-butadiene) features extended chemicals and chemotherapy drugs splash protection without compromising comfort
- Beaded cuffs and textured fingertips enable excellent handling of both wet and dry materials
- Extensively tested under stringent conditions, gloves meet AQL 0.65 for water tightness which results in a low risk of pinholes
- Recyclable through The RightCycle™ Programme where available
Powder-free, not made with latex or silicone, Kimtech™ Polaris™ Nitrile Gloves are designed to keep hands comfortable and protected whilst ensuring that research applications can be carried out contamination-free. Designated as PPE Cat III according to Regulation (EU) 2016/425 and to the Regulation 2016/425 as brought into UK law and amended and are ideal for use in higher-risk applications, as well as being food contact approved.
Certifications: Assured Compliance: - PPE Cat III according to Regulation (EU) 2016/425 and the Regulation 2016/425 as brought into UK law and amended; - EN ISO 374-1 Type B (JKOPT) Chemical Splash protection; - EN 374-4 Resistance to degradation by chemicals; - EN ISO 374-5 Micro Organism and VIRUS Protection; - ASTM D6978 Chemotherapy Drug Splash Protection; ISO 10993 Bio-compatibility; - Food contact approved.
Request a Quote
Description | UOM | VWR Catalog Number |
GLOVE KIMTECH POLARIS NITRILE 24CM XS | 1 * 100 items | KIMB62000 |
GLOVE KIMTECH POLARIS NITRILE 24CM S | 1 * 100 items | KIMB62001 |
GLOVE KIMTECH POLARIS NITRILE 24CM M | 1 * 100 items | KIMB62002 |
GLOVE KIMTECH POLARIS NITRILE 24CM L | 1 * 100 items | KIMB62003 |
GLOVE KIMTECH POLARIS NITRILE 24CM XL | 1 * 90 items | KIMB62004 |
GLOVE KIMTECH POLARIS XTRA NITR. 30CM XS | 1 * 50 items | KIMB62100 |
GLOVE KIMTECH POLARIS XTRA NITR. 30CM S | 1 * 50 items | KIMB62101 |
GLOVE KIMTECH POLARIS XTRA NITR. 30CM M | 1 * 50 items | KIMB62102 |
GLOVE KIMTECH POLARIS XTRA NITR. 30CM L | 1 * 50 items | KIMB62103 |
GLOVE KIMTECH POLARIS XTRA NITR. 30CM XL | 1 * 50 items | KIMB62104 |
*Terms and Conditions:
- Promotion valid from now till 30 June 2024 *While Stocks Last.
- VWR standard Terms and Conditions apply
- All pre-orders must be made within the promotional period.
- KC Polaris Gloves New Launch is not applicable with other on-going promotions.