VWR® Nano Spectrophotometer
VWR® Nano Spectrophotometer is designed to simplify and accelerate your molecular biology research and routine applications. The VWR® Nano Spectrophotometer allows for the swift and accurate purity analysis and quantification of nucleic acids and proteins with as little as 0,5 µL of sample.
Up to 37% OFF! While stocks last. *T&Cs Apply.
- Rapid calculation and display of 260/280 and 260/230 absorbance ratios
- Extended-life xenon lamp and monochromator for full spectral scan
- Auto adjusting pathlengths: 0,2 and 1,0 mm, ensuring accurate measurements
- Detects up to 4500 ng/µl of undiluted dsDNA
- Built-in printer and USB port
The VWR® Nano Spectrophotometer allows for the swift and accurate purity analysis and quantification of nucleic acids and proteins with as little as 0,5 µL of sample. This stand-alone device operates through a full-colour, 17,8 cm touchscreen with preset programs for nucleic acids and protein sample evaluation, colorimetric assays (BCA, Bradford, Lowry, etc.), UV-Vis spectral scanning, and OD 600 measurements to determine bacteria culture growth. The on-board software includes a useful 'Protocol Guidance' feature to guide the user through each step of the process.
At the heart of the device is a long-life xenon lamp which provides a wide wavelength range from 200 to 800 nm. The xenon lamp system does not require any warm-up time; the instrument can be used immediately upon power-up. A highly sensitive, high-speed silicon CCD detector ensures fast and accurate results.
Certifications: CE and UKCA marked.
Delivery information: Supplied with software, EU plug and UK plug, and 2 glass cuvettes.
Request a QuoteDescription | UOM | VWR Catalog Number |
VWR NANO SPECTROPHOTOMETER EU/UK PLUG | 1 * 1 items | VWRI76777-152 |
*Terms and Conditions:
- Promotion valid from now till 31 December 2024 *While Stocks Last.
- VWR standard Terms and Conditions apply
- All pre-orders must be made within the promotional period.
- VWR Nano Spectrophotometer is not applicable with other on-going promotions.