VWR Cell Counters
Newly Launched! Enjoy 20% off with Avantor VWR. While stocks last. *T&Cs Apply.
Save time and reduce variability with VWR® automated cell counter and VWR® automated cell counter Fluo. Intelligent image recognition and advanced optical imaging technology to achieve accurate and automatic cell analysis, cell concentration and cell size determination.
- Wide application: cell counting, cell viability, AO/PI staining, cell transfection efficiency, cell expression
- All-in-one machine with a compact design
- Dual channel option in VWR® automated cell counter Fluo
VWR® automated cell counter Fluo dual channel: detachable building block design enables easy installation and replacement of the fluorescent cubes. Fluorescent cubes sold separately. FL Cube blue for DAPI, Hoechst, BFP FL cube green for acridine orange(AO), Calcein-AM, Alexa fluor 488, SYBR green, FITC, GFP FL cube red for propidium iodide(PI), DsRed, dTomato, RFP.
Certifications: CE and UKCA marked.
Description | UOM | VWR Catalog Number |
VWR® automated cell | 1 * 1 items | VWRI49862-2000 |
VWR® Fluo cube for DAPI, blue | 1 * 1 items | VWRI49893-4950 |
VWR® Fluo cube for GFP, green | 1 * 1 items | VWRI49893-4951 |
VWR® Fluo cube for PI, red | 1 * 1 items | VWRI49893-4952 |
VWR® automated cell counter Fluo | 1 * 1 items | VWRI49893-2000 |
VWR® cell counting slides | 1 * 50 items | VWRI10228-0050 |
*Terms and Conditions:
- Promotion valid from now till 31 December 2023 *While Stocks Last.
- VWR standard Terms and Conditions apply
- All pre-orders must be made within the promotional period.
- VWR Cell Counters is not applicable with other on-going promotions.