Avantor Day Exclusive 2024 - Pipettes
Precision in Every Drop: Get 5 Pipettes for Just 100 SGD Each! While stocks last. *T&Cs Apply.
For every 500 SGD worth of chemical purchase order, the customer is entitled to buy a maximum of 5 pipettes at 100 SGD each.
VWR PHP Pipettes are fully autoclavable, high performance pipettes with low plunger force.
- Superior accuracy & precision (tested in ISO 17025 accredited lab)
- Magnet assisted piston for low plunger force and lesser chance of RSI
- PVDF tip cone for durable performance
- Large and clear 4 digit display; color coding for easy identification
- 3-Year warranty
VWR PHP Pipettes are Precise High Performance, fully autoclavable pipettes, that are available in Single and Multi Channel range. VWR PHP offers one of the lowest plunger forces and a durable tip cone. The tip ejector is made using special corrosion resistant alloy, protecting it against chemical and physical corrosion. The pipette has an effortless tip ejection with unique shock absorbing mechanism. The tip cone is spring loaded for lesser strain and proper tip fitment for precise pipetting (for Multi Channel only).
Description | UOM | VWR Catalog Number |
PHP - SINGLE CH VV PIPETTE 0.1-2 | 1 * 1 items | VWRA613-6688 |
PHP - SINGLE CH VV PIPETTE 0.5-10 | 1 * 1 items | VWRA613-6689 |
PHP - SINGLE CH VV PIPETTE 2-20 | 1 * 1 items | VWRA613-6690 |
PHP - SINGLE CHANNEL VV PIPETTE 5-50 | 1 * 1 items | VWRA613-6691 |
PHP - SINGLE CH VV PIPETTE 10-100 | 1 * 1 items | VWRA613-6692 |
PHP - SINGLE CH VV PIPETTE 20-200 | 1 * 1 items | VWRA613-6693 |
PHP - SINGLE CH VV PIPETTE 100-1000 | 1 * 1 items | VWRA613-6694 |
PHP - SINGLE CH VV PIPETTE 1000-5000 | 1 * 1 items | VWRA613-6695 |
PHP - SINGLE CH VV PIPETTE 1000-10000 | 1 * 1 items | VWRA613-6696 |
*Terms and Conditions:
- Promotion valid from now till 31 March 2024 *While Stocks Last.
- VWR standard Terms and Conditions apply
- All pre-orders must be made within the promotional period.
- For every 500 SGD worth of chemical purchase order, the customer is entitled to buy a maximum of 5 pipettes at 100 SGD each.